missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FN3KRP Polyclonal Antibody
GREENER_CHOICE

Código de producto. 13422257
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
13422257 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 13422257

Marca: Invitrogen™ PA528166

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (86%), Rat (86%), Dog (84%), Rhesus Monkey (82%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Fructosamine-3-kinase related protein (FN3KRP) is located on chromosome 17q25.3, approximately 8 kb upstream of the FN3K gene. It shares 65% amino acid identity with FN3K and possesses an identical genomic organization, implying functional similarities; FN3KRP is suggested to play a role in enzymatic deglycation. Unlike FN3K, FN3KRP is more ancient, present in diverse organisms such as cyanobacteria, echinoderms, plants, and cold-blooded vertebrates. Studies have discovered a significant association between rs1046896 in FN3KRP and longevity, where the longevity allele C is linked to increased FN3KRP expression in whole blood. FN3KRP is hypothesized to phosphorylate glucitolamines, products of non-enzymatic glycation, indicating its involvement in crucial metabolic processes. The regulation of both FN3K and FN3KRP is notable for TATA-less and CAAT-less promoters, featuring homologous CpG islands and Sp1 binding sites. Meanwhile, both genes are widely expressed across human tissues, but FN3K demonstrates significantly higher levels in tissues prone to glycation-related complications.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno FN3KRP
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.57 mg/mL
Conjugado Unconjugated
Formulación 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
génica FN3KRP
N.º de referencia del gen Q9HA64
Alias de gen FN3KL; FN3K-related protein; Fn3krp; FN3K-RP; fructosamine 3 kinase related protein; fructosamine 3-kinase-related protein; fructosamine-3-kinase-related protein; Ketosamine-3-kinase; ortholog of human fructosamine-3-kinase-related protein FN3KRP; Protein-psicosamine 3-kinase FN3KRP; RGD1304570; testis secretory sperm-binding protein Li 211a
Símbolos de los genes FN3KRP
Especie del huésped Rabbit
Inmunógeno Recombinant protein encompassing a sequence within the center region of human FN3KRP. The exact sequence is proprietary.
Método de purificación Antigen affinity chromatography
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79672
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.