missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FMO5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA551185
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The antibody detects endogenous levels of total FMO5 protein.
Metabolic N-oxidation of the diet-derived amino-trimethylamine (TMA) is mediated by flavin-containing monooxygenase and is subject to an inherited FMO3 polymorphism in man resulting in a small subpopulation with reduced TMA N-oxidation capacity resulting in fish odor syndrome Trimethylaminuria. Three forms of the enzyme, FMO1 found in fetal liver, FMO2 found in adult liver, and FMO3 are encoded by genes clustered in the 1q23-q25 region. Flavin-containing monooxygenases are NADPH-dependent flavoenzymes that catalyzes the oxidation of soft nucleophilic heteroatom centers in drugs, pesticides, and xenobiotics. Alternative splicing results in multiple transcript variants.
Especificaciones
| FMO5 | |
| Polyclonal | |
| Unconjugated | |
| FMO5 | |
| 5033418D19Rik; AI195026; Dimethylaniline monooxygenase [N-oxide-forming] 5; dimethylaniline oxidase 5; flavin containing monooxygenase 5; FMO 5; Fmo5; Hepatic flavin-containing monooxygenase 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14263, 2330 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P49326, P97872 | |
| FMO5 | |
| Fusion protein of human FMO5. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido