missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMO2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA555810
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: IFPTAELQAR WVTRVFKGLC SLPSERTMMM DIIKRNEKRI DLFGESQSQT LQTNYVDYLD ELALEIGAKP DFCSLLFKDP KLAVRLYFGP CN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
The flavin-containing monooxygenases are NADPH-dependent enzymes that catalyze the oxidation of many drugs and xenobiotics. In most mammals, there is a flavin-containing monooxygenase that catalyzes the N-oxidation of some primary alkylamines through an N-hydroxylamine intermediate. However, in humans, this enzyme is truncated and is probably rapidly degraded. The protein encoded by this gene represents the truncated form and apparently has no catalytic activity. A functional allele found in African Americans has been reported, but no sequence evidence has been deposited to support the finding. This gene is found in a cluster with the FMO1, FMO3, and FMO4 genes on chromosome 1.
Especificaciones
| FMO2 | |
| Polyclonal | |
| Unconjugated | |
| FMO2 | |
| FMO1B1, FMO 1B1, FMO 2, FMO, pulmonary, dimethylaniline monooxygenase [N-oxide-forming] 2, dimethylaniline oxidase 2, pulmonary flavin-containing monooxygenase 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2327, 55990 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99518, Q8K2I3 | |
| FMO2 | |
| Recombit protein corresponding to Human FMO2 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido