missing translation for 'onlineSavingsMsg'
Learn More

FLYWCH2 Rabbit, anti-Human, Polyclonal, Proteintech

Product Code. 18047045 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
Product Code. Cantidad unitSize
18047045 20 μL 20 microlitros
18068715 150 μL 150 microlitros
2 options
This item is not returnable. View return policy

Product Code. 18047045

Brand: Proteintech 242281AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The FLYWCH2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FLYWCH2. This antibody recognizes human antigen. The FLYWCH2 antibody has been validated for the following applications: IHC, ELISA analysis.

Specifications

Antígeno FLYWCH2
Aplicaciones Immunohistochemistry, ELISA
Concentración 700 μg/mL
Símbolos de los genes FLYWCH family member 2, FLYWCH2
Especie del huésped Rabbit
Cantidad 20 μL
Primario o secundario Primary
ID de gen (Entrez) 114984
Especies diana Human
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 150 kDa
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.