missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FLYWCH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA559385
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: TLASTLQILP VEEQGGVVQP ALEMPEQKCS KLDAAAPQSL EFLRTPFGGR LLVLESFLYK QEKAVGDKVY WKCRQHAELG CRGRAITRGL RATVMRGHC Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 65%.
Transcription cofactor. Negatively regulates transcription activation by catenin beta-1 CTNNB1, perhaps acting by competing with TCF4 for CTNNB1 binding. May play a role in DNA-damage response signaling. Binds specifically to DNA sequences at peri-centromeric chromatin loci.
Especificaciones
| FLYWCH1 | |
| Polyclonal | |
| Unconjugated | |
| FLYWCH1 | |
| E030034P13Rik; FLYWCH1; FLYWCH-type zinc finger 1; FLYWCH-type zinc finger-containing protein 1; KIAA1552; mKIAA1552 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84256 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4VC44 | |
| FLYWCH1 | |
| Recombinant protein corresponding to Human FLYWCH1. Recombinant protein control fragment (Product #RP-98607). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido