Learn More
Invitrogen™ Fluorescein/Oregon Green Polyclonal Antibody, Alexa Fluor™ 488
Descripción
We offer a variety of anti-fluorescent dye antibodies that recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria, or other biomolecules have been internalized by endocytic or pinocytic processes. In addition to their utility in quenching the fluorescence of extracellular fluorescein, the high affinity of Invitrogen's anti-fluorescein antibodies for both fluorescein and the structurally related Oregon GreenTM dye makes them ideal for immunochemical applications. Harmer and Samuel report the fluorescein-anti-fluorescein system provides a sensitive alternative to the biotin-streptavidin methods conventionally used for enzyme-linked immunosorbent assays (ELISAs). Fluorescein has also been found to be an excellent hapten for FISH, yielding sensitive results with extremely low background levels. The Alexa Fluor 488 conjugate can be used to amplify the fluorescence signal while still allowing fluorescein-compatible optics. The approximate absorption (Abs) and fluorescence emission (Em) maxima in nm: 495/519. Our anti-dye antibodies can be used in many different applications. It is a good practice to centrifuge the labeled antibody solutions briefly in a microcentrifuge before use; only the supernatant should then be added to the experiment. This step will eliminate any...
Especificaciones
Especificaciones
| Antígeno | Fluorescein/Oregon Green |
| Aplicaciones | ELISA |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Alexa Fluor 488 |
| Formulación | 0.1M potassium phosphate with 5mM sodium azide; pH 8 |
| Alias de gen | FITC |
| Especie del huésped | Goat |
| Inmunógeno | Fluorescein/Oregon Green. |
| Método de purificación | Purified |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.