Learn More
Invitrogen™ Fluorescein/Oregon Green Polyclonal Antibody, Alexa Fluor™ 488
GREENER_CHOICE
Código de producto. 10572853
Change view
Click to view available options
Cantidad:
500 μL
Tamaño de la unidad:
cada uno
10572853 500 μL cada uno
1 options
Código de producto. 10572853 Proveedor Invitrogen™ N.º de proveedor A11090
Rabbit Polyclonal Antibody
The Alexa Fluor 488 conjugates can be used to amplify the fluorescence signal while still allowing fluorescein-compatible optics. Excitation 495 nm/peak emission 519 nm. It is a good practice to centrifuge the labeled antibody solutions briefly in a microcentrifuge before use; only the supernatant should then be added to the experiment. This step will eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
Anti-fluorescent dye antibodies recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria or other biomolecules have been internalized by endocytic or pinocytic processes.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Fluorescein/Oregon Green
Aplicaciones ELISA, Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Alexa Fluor 488
Formulación 0.1M potassium phosphate with 5mM sodium azide; pH 8
Alias de gen FITC
Especie del huésped Rabbit
Inmunógeno Fluorescein/Oregon Green.
Método de purificación Purified
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
Especies diana Chemical
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Molécula diana Chemical