Learn More
Cole-Parmer Flow Through Tubing Weight, PTFE
Código de producto. p-200105083 Tienda Cole-Parmer Productos
Change view
Click to view available options
Tamaño de los tubos:
De 3/16 in a 1/4 in
De 5/16 in a 7/16 in
Tamaño de la unidad:
cada uno
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Tamaño de los tubos unitSize
30053089 De 3/16 in a 1/4 in cada uno
30054041 De 5/16 in a 7/16 in cada uno
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30053089 Proveedor Cole-Parmer N.º de proveedor 5011798
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Prevent tubing from coiling or floating during fluid transfer applications
Tapered hose barb fitting easily inserts into the inner diameter (ID) of flexible tubing. Keep tubing submerged in liquid, preventing floatation, and ensuring uninterrupted fluid flow. Constructed of chemically inert PTFE. Autoclavable—allowing for easy sterilization and reuse
Especificaciones
Tamaño de los tubos De 3/16 in a 1/4 in