Learn More
Proteintech FlexAble HRP Antibody Labeling Kit for Human IgG
Código de producto. p-200050243 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
10 pruebas
100 pruebas
50 pruebas
Tamaño de la unidad:
10 reacciones
200 reacciones
50 reacciones
18046195 10 pruebas 10 reacciones
18025595 50 pruebas 50 reacciones
18060336 100 pruebas 200 reacciones
3 options
Código de producto. 18046195
Marca: Proteintech KFA11010
FlexAble HRP Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate HRP in any buffer condition to Human IgG primary antibodies from any supplier.
FlexAble HRP Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate HRP in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Especificaciones
Cantidad 10 pruebas
Formato Liquid