Learn More
Fisherbrand™ Vial Screw Caps
Fisherbrand™ Vial Screw Caps are for externally threaded sample tubes and feature long skirts and a super-fast thread design that allows them to be removed or sealed with single turn.
Descripción
Descripción
  • Made of polypropylene
  • With lip seal design for leakproof seal
  • Autoclavable
  • For externally threaded tubes
  • Tubes sold separately