Description
Made of plastic for use on standard taper glass joints
- Color coded for easy size identification
- Does not corrode
- Will not scratch glass
- Withstands temperatures up to 150°C
Specifications
Specifications
| Cantidad | 10/Pk. |
| Tipo | Clip de unión |
| Material | Plastic |
| Tamaño | Tamaño de la junta: 29 |
| Color | Rojo |