missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ Digital and Analog Dry Bath Incubators

Código de producto. p-3275597 Tienda Fisherbrand Productos
Change view
Click to view available options
Tipo de controlador:
Digital
N.º de bloques:
4
Tamaño de la unidad:
cada uno
Código de producto. Tipo de controlador N.º de bloques unitSize
11830740 Digital 4 cada uno
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11830740

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Includes

Digital Models come with temperature probe, line cord and plug. Analog Models include high and low thermostat, power switch, pilot light, line cord and plug.

Requires

Heating blocks, sold separately, hold regular tubes, microcentrifuge tubes, square cuvets, or microplates.

Provide controlled dry heat for a wide variety of clinical and general chemistry applications

Choice of 1- to 6-block Analog Models, 2- or 4-block Digital Models.

  • Exceptional temperature control and uniformity
  • Temperature range of 5°C above ambient to 130°C
  • Powder-coated steel body construction provides durability for demanding applications
  • Compact design saves valuable bench space
  • Choice of interchangeable heating blocks

Digital Dry Baths with Microprocessor Controller

  • PID microprocessor control, digital temperature setting
  • Just press up/down arrows to set temperature in 0.1° increments
  • Effectively hold samples at constant, warm temperature
  • 5°C above ambient to 130°C range; ±0.4° uniformity at 37°C; stability, ±0.2°C
  • Bright LED display continuously shows actual temperature in °C
  • Temperature probe in bottom of bath provides improved accuracy and control; free probe included
  • Model 125DQ holds two heating blocks; Model 145DQ holds four heating blocks in 2 × 2 configuration
Analog Dry Baths with Dual-thermostat Controls
  • Sloped control panel makes controls highly visible
  • Dual bimetallic thermostat controls
  • Stability is ±1.5°C, uniformity ±0.5°C at 37°C
  • Use thermometer to read current temperature of blocks, then adjust control to reach desired setpoint
  • Efficient design: operate at substantially lower external temperature than other models

Applications:

Ideal for enzyme reactions, inactivation of sera, Rh studies, blood crossmatching and cholesterol determinations

Información de órdenes

All models accept 2.91L x 3.75W x 2 in.H (7.4 x 9.5 x 5.1cm) blocks, sold separately.

Certifications

Digital Models are CSA approved. Analog Models are UL listed.

Garantía

Un año

†International model

TRUSTED_SUSTAINABILITY

Especificaciones

Tipo de controlador Digital
Material Acero
Intervalo de temperatura (métrico) Ambient +5° to 130°C
Incluye Microprocessor controller, temperature probe, line cord and plug, holds two heating blocks
N.º de bloques 4
Dimensiones (Largo x Ancho x Alto) 14.2 x 10.8 x 3.3 in. (36.1 x 27.4 x 9.1 cm)
Longitud (imperial) 14.2 in.
Longitud (métrico) 36.1 cm
Anchura (imperial) 10.8 in.
Anchura (métrico) 27.4 cm
Altura (imperial) 3.3 in.
Altura (métrico) 9.1 cm
Requisitos eléctricos 240 V, 50/60 Hz
Tensión 240 V
Frecuencia 50/60 Hz
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.