missing translation for 'onlineSavingsMsg'
Learn More

Fimasartan, MedChemExpress

Product Code. 16461058
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
50 mg
100 mg
Unit Size:
10 miligramos
100 miligramos
5 miligramos
50 miligramos
Product Code. Cantidad unitSize
16461058 5 mg 5 miligramos
16468608 10 mg 10 miligramos
16448456 100 mg 100 miligramos
16414487 50 mg 50 miligramos
4 options
This item is not returnable. View return policy

Product Code. 16461058

Brand: Medchem Express HYB0780

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Fimasartan(BR-A-657) is a non-peptide angiotensin II receptor antagonist used for the treatment of hypertension and heart failure.

Fimasartan(BR-A-657) is a non-peptide angiotensin II receptor antagonist used for the treatment of hypertension and heart failure.

Specifications

Nombre del producto químico o material Fimasartan
Forma física Solid
CAS 247257-48-3
Color Amarillo claro
Cantidad 5 mg
Notas de grado de pureza Research
Fórmula molecular C27H31N7OS
Para utilizar con (aplicación) COVID-19-immunoregulation
Sinónimo BR-A-657
Información de solubilidad DMSO : ≥ 49 mg/mL (97.68 mM)
SMILES O=C1N(C(CCCC)=NC(C)=C1CC(N(C)C)=S)CC2=CC=C(C3=C(C4=NN=NN4)C=CC=C3)C=C2
Peso molecular (g/mol) 501.65
Formula Weight (peso de la fórmula) 501.65
Porcentaje de pureza 98.04%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.