Learn More
CELLPATH FILOSLIDE 50 BOX
Click to view available options
Color:
Black
Blue
White
Tamaño de la unidad:
cada uno
Descripción
- Key Benefits:
- Stackable
- Snap closure to seal out dust
- Splash-proof
- Double hinged lid prevents loss or breakage
- Index card included for easy specimen identification
- Can store up to 50 micrscope slides
Especificaciones
Especificaciones
| Tipo | Slide Box |
| Capacidad para portaobjetos | 50 |
| Color | Blue |