Learn More
CELLPATH FILOSLIDE 25 BOX
Descripción
- Key Benefits:
- Stackable
- Snap closure to seal out dust
- Splash-proof
- Double hinged lid prevents loss or breakage
- Index card included for easy specimen identification
- Can store up to 25 micrscope slides
Especificaciones
Especificaciones
| Tipo | Slide Box |
| Capacidad para portaobjetos | 25 |
| Color | Red |