missing translation for 'onlineSavingsMsg'
Learn More

FIGF, Mouse anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16138154
Change view
Click to view available options
Cantidad:
50 μg
Unit Size:
50 microgramos
Product Code. Cantidad unitSize
16138154 50 μg 50 microgramos
1 options
This item is not returnable. View return policy

Product Code. 16138154

Brand: Abnova H00002277B01P.50ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Sequence: MYREWVVVNVFMMLYVQLVQGSSNEHGPVKRSSQSTLERSEQQIRAASSLEELLRITHSEDWKLWRCRLRLKSFTSMDSRSASHRSTRFAATFYDIETLKVIDEEWQRTQCSPRETCVEVASELGKSTNTFFKPPCVNVFRCGGCCNEESLICMNTSTSYISKQLFEISVPLTSVPELVPVKVANHTGCKCLPTAPRHPYSIIRRSIQIPEEDRCSHSKKLCPIDMLWDSNKCKCVLQEENPLAGTEDHSHLQEPALCGPHMMFDEDRCECVCKTPCPKDLIQHPKNCSCFECKESLETCCQKHKLFHPDTCSCEDRCPFHTRPCASGKTACAKHCRFPKEKRAAQGPHSRKNP

Specifications

Antígeno FIGF
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Formulación In 1x PBS, pH 7.4
génica c-fos induced growth factor (vascular endothelial growth factor D)
N.º de referencia del gen NM_004469
Alias de gen VEGF-D/VEGFD
Símbolos de los genes FIGF
Especie del huésped Mouse
Inmunógeno FIGF (NP_004460, 1 a.a. ∼ 354 a.a) full-length human protein.
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2277
Especies diana Human
Contenido y almacenamiento Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Tipo de producto Antibody
Formulario Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.