missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrillarin Monoclonal Antibody (38F3)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA316771
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Suggested positive control: antigen standard for FBL (transient overexpression lysate).
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).
Especificaciones
| Fibrillarin | |
| Monoclonal | |
| Conc. not determined | |
| Tissue culture supernatant with 0.035% sodium azide | |
| P15646, P22087, P22509, P35550, Q22053 | |
| fbl, fib-1, Nop1p | |
| Yeast nuclear preparation (S. cerevisiae). | |
| RUO | |
| 100064571, 100626002, 14113, 179999, 2091, 292747, 46234, 538548, 851548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 38F3 | |
| Unconjugated | |
| fbl | |
| 34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7 | |
| Mouse | |
| 250 μL | |
| Primary | |
| Bovine, C. elegans, Chicken, Drosophila, Horse, Human, Mouse, Plant, Pig, Rat, Yeast | |
| Antibody | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto