Learn More
FIBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Código de producto. 16875384 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16875384 20 μL 20 microlitros
16865384 150 μL 150 microlitros
2 options
Código de producto. 16875384
Marca: Proteintech 159681AP20UL
Rabbit Polyclonal Antibody
Acidic fibroblast growth factor is mitogenic for a variety of different cell types and acts by stimulating mitogenesis or inducing morphological changes and differentiation. The FIBP protein is an intracellular protein that binds selectively to acidic fibroblast growth factor (aFGF). It is postulated that FIBP may be involved in the mitogenic action of aFGF. Two transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Antígeno FIBP
Aplicaciones Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Clasificación Polyclonal
Concentración 0.17 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica FIBP
N.º de referencia del gen O43427, Q9JI19
Alias de gen FGFIBP, FIBP, FIBP 1
Símbolos de los genes Fibp
Especie del huésped Rabbit
Inmunógeno FIBP Fusion Protein Ag8743
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 58249, 9158
Especies diana Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG