missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FIBB Monoclonal Antibody (1F9)

Descripción
MA5-15519 targets FGB in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15519 immunogen is purified recombinant fragment of human FGB (aa30-300) expressed in E. Coli. MA5-15519 detects FGB which has a predicted molecular weight of approximately 52kDa.
FIBB is the beta component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in its gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.
Especificaciones
Especificaciones
| Antígeno | FIBB |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 1F9 |
| Concentración | Conc. Not Determined |
| Conjugado | Unconjugated |
| Formulación | Ascites with 0.03% sodium azide |
| génica | Fgb |
| N.º de referencia del gen | P02675 |
| Alias de gen | 2510049G14Rik; Ab1-181; Ab1-216; Ac1-581; B beta polypeptide; beta-fibrinogen; epididymis secretory sperm binding protein Li 78p; Fgb; fibrinogen; Fibrinogen beta chain; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; Fibrinopeptide B; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC104327; MGC120405 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?