Learn More
Invitrogen™ Fianlimab Recombinant Monoclonal Antibody
Descripción
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Antibody pharmaceuticals against LAG-3 are indicated for malignant tumors.
Especificaciones
Especificaciones
| Antígeno | Fianlimab |
| Aplicaciones | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Clasificación | Recombinant Monoclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 |
| Alias de gen | REGN3767 |
| Especie del huésped | Human |
| Método de purificación | Protein A |
| Cantidad | 100 μg |
| Mostrar más |