missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF17 Polyclonal Antibody

Descripción
Immunogen sequence: TQGENHPSPN FNQYVRDQGA MTDQLSRRQI REYQLYSRTS GKHVQVTGRR ISATAEDGNK FAKLIVETDT FGSRVRIKGA ESEKYICMNK RGKLIGKPSG KSKDCVFTEI VLENNYTAFQ NARHEGWFMA FTRQGRPRQA SRSRQNQREA HFIKRLYQ.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17.
Especificaciones
Especificaciones
| Antígeno | FGF17 |
| Aplicaciones | ELISA, Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1.46 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| génica | FGF17 |
| N.º de referencia del gen | O60258, P63075, P63076 |
| Alias de gen | FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187 |
| Símbolos de los genes | FGF17 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?