missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: EPLLDTHIVN GERDETATAP ASPTTDSCDG NASDSSYRTP GIGPVLPLEE RGAETETKVQ ERENGESPLE LEQLDQHHEM KETNEQ Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 52%.
The FGD4 gene, also known as FYVE, RhoGEF, and PH domain-containing protein 4, is located on chromosome 12p11.21. This gene encodes a protein involved in the regulation of the actin cytoskeleton and is crucial for cellular processes such as migration and adhesion. FGD4 is specifically associated with Charcot-Marie-Tooth disease type 4H (CMT4H), a rare autosomal recessive hereditary neuropathy characterized by early onset, progressive distal muscle weakness, scoliosis, and myelin outfoldings. Mutations in FGD4 result in disruptions of its function, leading to the clinical manifestations of CMT4H. Additionally, the gene has been implicated in cancer biology, with its expression correlating with aggressive phenotypes in some cancers and contributing to therapeutic resistances. The gene includes multiple exons and alternative splicing variants, underlining its broad functional implications.
Especificaciones
Especificaciones
| Antígeno | FGD4 |
| Aplicaciones | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 0.05 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | FGD4 |
| N.º de referencia del gen | Q96M96 |
| Alias de gen | Actin filament-binding protein frabin; actin-filament binding protein frabin; CMT4H; FGD1 family, member 4; FGD1-related F-actin-binding protein; FGD4; FRABP; FYVE, RhoGEF and PH domain containing 4; FYVE, RhoGEF and PH domain-containing protein 4; ZFYVE6; Zinc finger FYVE domain-containing protein 6 |
| Símbolos de los genes | FGD4 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
