missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FFAR3/GPR41 Antibody (2590G) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals FAB10562JF669
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FFAR3/GPR41 Monoclonal antibody specifically detects FFAR3/GPR41 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Especificaciones
| FFAR3/GPR41 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| free fatty acid receptor 3, G protein-coupled receptor 41, GPR41FFA3R, G-protein coupled receptor 41 | |
| Synthetic peptide containing human FFAR3 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 2590G | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| GPCR | |
| 2865 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction