missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fetal Hemoglobin Monoclonal Antibody (HBF-1), FITC, Invitrogen™
GREENER_CHOICE

Código de producto. p-7111509
Change view
Click to view available options
Cantidad:
500 μL
2 mL
Tamaño de la unidad:
cada uno
Código de producto. Cantidad unitSize
10716293 500 μL cada uno
10632024 2 mL cada uno
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 10716293

Marca: Invitrogen™ MHFH01

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility. This Fetal Hemoglobin monoclonal antibody specifically recognizes fetal hemoglobin found in fetal erythrocytes.

Hemorrhage of cells from an Rh+ fetus into the circulation of an Rh- mother may result in the formation of Rh-reactive antibodies in the mother. Rh hemolytic anemia in a subsequent Rh+ fetus may result from placental transfer of antibodies formed in the mother to the fetus. Although the volume of fetal erythrocytes found in the maternal circulation during pregnancy and immediately post-partum is reported to be small and of uncertain clinical significance in many cases, substantial hemorrhage may result from a number of causes including fetal or maternal trauma and placental defects. Erythrocytes containing fetal hemoglobin may be found in individuals of any age, but with lower amounts of fetal hemoglobin compared to fetal red cells. These cells have been termed F cells. High levels of F cells may also exist in adults with a heterogeneous group of genetic disorders of uncertain etiology, referred to as Hereditary Persistence of Fetal Hemoglobin. Other clinical conditions causing significant levels of anemia may also result in elevated levels of F cells. Several clinical conditions have been described with increased levels of F cells. These conditions include hereditary anemic diseases such as sickle cell anemia and thalassemia major.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Fetal Hemoglobin, Fetal Hemoglobin (HbF)
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon HBF-1
Conjugado FITC
Formulación PBS with BSA, <10% sucrose and 0.1% sodium azide
génica HBG1
N.º de referencia del gen P69891
Alias de gen A-gamma globin; alpha-1 globin; gamma A hemoglobin; gamma globin; gamma-1-globin; Hb F Agamma; HBG1; HBGA; HBGR; HBG-T2; hemoglobin alpha 1; Hemoglobin gamma-1 chain; hemoglobin gamma-a chain; hemoglobin subunit gamma 1; hemoglobin subunit gamma-1; hemoglobin, gamma A; hemoglobin, gamma, regulator of; HSGGL1; PRO2979
Símbolos de los genes HBG1
Especie del huésped Mouse
Inmunógeno Human Fetal Hemoglobin (HbF).
Método de purificación Purified
Cantidad 500 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3047
Especies diana Human
Contenido y almacenamiento 4°C, store in dark
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Características ecológicas Responsibly packaged
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.