Learn More
Value Plastics Female Luer Thread Style to NPT Adapter Fittings, Straight
Tienda Cole-Parmer ProductosDescripción
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Especificaciones
Especificaciones
| Certificaciones/conformidad | Non-DEHP;Non-Phthalate;RoHS |
| Conexión | 1/8-27 NPT(M) Thread |
| Material (sello) | Nailon blanco |