Descripción
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Especificaciones
Especificaciones
| Certificaciones/conformidad | Non-DEHP;Non-Phthalate;RoHS;FDA |
| Diámetro interior (imperial) | 1/16 in |
| Material | Policarbonato |
| Cantidad de embalaje | 25/Pk. |
| Conexión | Female Luer with Panel Mount, Hose Barb |
Always test the application to verify the performance of the interconnection.