Learn More
Value Plastics Female Luer to 400 Series Barb
Código de producto. 30053500 Tienda Cole-Parmer Productos
Click to view available options
Diámetro interior (imperial):
1/16 in
1/8 in
Tamaño de la unidad:
paquete de 25
Código de producto. 30053500
Marca: Value Plastics 5012056
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Especificaciones
Certificaciones/conformidad ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diámetro interior (imperial) 1/16 in
Material Polipropileno
Cantidad de embalaje 25/Pk.
Conexión Female Luer x Hose Barb
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.