missing translation for 'onlineSavingsMsg'
Learn More

FCRN/FCGRT Antibody (nipocalimab) [Janelia Fluor™ 635], Novus Biologicals™

Product Code. 30160984
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30160984 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30160984

Brand: Novus Biologicals (Bio-Techne) NBP328599JF635

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Human Monoclonal Antibody

FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antígeno FCRN/FCGRT
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon nipocalimab
Conjugado Janelia Fluor 635
Formulación 50mM Sodium Borate
Alias de gen alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
Especie del huésped Human
Inmunógeno FcRn (FCGRT & B2M)
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 2217
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.