missing translation for 'onlineSavingsMsg'
Learn More

FCGR3A Monoclonal Antibody (OTI12H8), TrueMAB™, OriGene

Código de producto. 17518364
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
17518364 30 μL 30 microlitros
17598354 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17518364 Supplier Origene Technologies Supplier No. TA813635S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.

FCGR3A Monoclonal antibody specifically detects FCGR3A in Human samples. It is validated for Flow Cytometry

Specifications

Antígeno FCGR3A
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon OTI12H8
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica FCGR3B
N.º de referencia del gen O75015, P08637
Alias de gen 4833442P21Rik; CD16; CD16-2; CD16A; CD16a antigen; CD16b; CD32; CD32 receptor 2; CDw32; cytolytic trigger molecule G7; Fc fragment of IgG intermediate affinity IV receptor; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIIa; Fc fragment of IgG receptor IIIb; Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor; Fc fragment of IgG, low affinity IIIa, receptor (CD16a); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor for (CD16); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIb, receptor (CD16b); Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor IIIb; Fc gamma RIIIa; Fc gammaRIV; Fc receptor, IgG, low affinity III; Fc receptor, IgG, low affinity IV; Fc receptor-like 3; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD 16);
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human FCGR3A (NP_001121065) produced in HEK293T cell.
Método de purificación Protein A/G
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2214, 2215
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.