missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody recognizes isoform 1 (45 kDa) and isoform 3 (30-33 kDa).
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Two transcript variants encoding different isoforms exist for this gene.
Specifications
Specifications
| Antígeno | FBXO22 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.17 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | FBXO22 |
| N.º de referencia del gen | Q8NEZ5 |
| Alias de gen | F box only protein 22, F box protein 22, F box protein FBX22p44, FBX22, FBXO22, FISTC1 |
| Símbolos de los genes | FBXO22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?