Learn More
FBXO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16803304 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16803304 20 μL 20 microlitros
16893294 150 μL 150 microlitros
2 options
Código de producto. 16803304
Marca: Proteintech 145901AP20UL
Rabbit Polyclonal Antibody
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.
Especificaciones
Antígeno FBXO2
Aplicaciones Western Blot, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 0.13 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica FBXO2
N.º de referencia del gen Q80UW2, Q9UK22
Alias de gen F box only protein 2, F box protein 2, FBG1, Fbs1, FBX2, FBXO2, NFB42
Símbolos de los genes Fbxo2
Especie del huésped Rabbit
Inmunógeno FBXO2 Fusion Protein Ag6122
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 230904, 26232, 85273
Especies diana Rat, Human, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG