Learn More
FATE1 Mouse anti-Human, Clone: OTI7B5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF505383
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a cancer-testis antigen that is highly expressed in hepatocellular carcinomas and other tumors and weakly expressed in normal tissues except testis. The protein is strongly expressed in spermatogonia, primary spermatocytes, and Sertoli cells in seminiferous tubules. This protein may have a role in the control of early testicular differentiation and cell proliferation.Spécification
| FATE1 | |
| Monoclonal | |
| Unconjugated | |
| FATE1 | |
| CT43, FATE | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 89885 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7B5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q969F0 | |
| FATE1 | |
| Full length human recombit protein of human FATE1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.