missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA551057
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FAP Polyclonal Antibody detects endogenous levels of FAP protein.
The protein encoded by this gene is a homodimeric integral membrane gelatinase belonging to the serine protease family. It is selectively expressed in reactive stromal fibroblasts of epithelial cancers, granulation tissue of healing wounds, and malignant cells of bone and soft tissue sarcomas. This protein is thought to be involved in the control of fibroblast growth or epithelial-mesenchymal interactions during development, tissue repair, and epithelial carcinogenesis.
Especificaciones
| FAP | |
| Polyclonal | |
| Unconjugated | |
| FAP | |
| 170 kDa melanoma membrane-bound gelatinase; Antiplasmin-cleaving enzyme FAP, soluble form; APCE; Dipeptidyl peptidase FAP; DPPIV; Fap; FAPA; FAPalpha; fibroblast activation protein; fibroblast activation protein alpha; fibroblast activation protein, alpha; gelatine degradation protease FAP; Integral membrane serine protease; post-proline cleaving enzyme; Prolyl endopeptidase FAP; seprase; Serine integral membrane protease; SIMP; Surface-expressed protease | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14089, 192203, 2191 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97321, Q12884 | |
| FAP | |
| Synthesized peptide derived from the Internal region of human Seprase. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido