missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM83H Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA555094
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: SDKDKCSAIF RSDSLGTQGR LSRTLPASAE ERDRLLRRME SMRKEKRVYS RFEVFCKKEE ASSPGAGEGP AEEGTRDSKV GKFVPKILGT F Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
The protein encoded by this gene plays an important role in the structural development and calcification of tooth enamel. Defects in this gene are a cause of amelogenesis imperfecta type 3 (AI3).
Especificaciones
| FAM83H | |
| Polyclonal | |
| Unconjugated | |
| FAM83H | |
| AA409316; AI3; FAM83H; FAM83H variant 1; FAM83H variant 2; family with sequence similarity 83 member H; family with sequence similarity 83, member H; FLJ46072; protein FAM83H; RGD1305866 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286077, 362937 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZRV2 | |
| FAM83H | |
| Recombinant protein corresponding to Human FAM83H. Recombinant protein control fragment (Product #RP-93935). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido