missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM3C Polyclonal Antibody

Descripción
Peptide sequence: DNWVFCGGKG IKTKSPFEQH IKNNKDTNKY EGWPEVVEME GCIPQKQD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 91%; Rat: 100%; Zebrafish: 86%.
FAM3C is a 227 amino acid secretory protein that belongs to a novel cytokine-like family with sequence similarity to FAM3 family. FAM3C is a signaling cytokine with a GG domain, hydrophobic leader sequence and an N-terminal signal peptide. FAM3C has a cytokine activity and shows a change in expression in pancreatic cancer-derived cells. It is ubiquitously expressed in most of the tissues. Reports suggest that FAM3C plays an important role in inner ear development and may thus be involved in cell differentiation and proliferation during inner ear embryogenesis.
Especificaciones
Especificaciones
| Antígeno | FAM3C |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | FAM3C |
| N.º de referencia del gen | Q92520 |
| Alias de gen | D6Wsu176e; FAM3 metabolism regulating signaling molecule C; Fam3c; fam3c protein; FAM3C-like protein; family with sequence similarity 3 member C; family with sequence similarity 3, member C; GS3786; Ilei; Interleukin-like EMT inducer; interleukin-like epithelial-mesenchymal transition inducer; predicted osteoblast protein; protein FAM3C; si:dkey-159a18.2; wu:fb99h11; wu:fi35h03; zgc:55444; zgc:77090 |
| Símbolos de los genes | FAM3C |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?