missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAM21A Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5111514
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: GIQAKTTKPK SRSAQAAPEP RFEHKVSNIF DDPLNAFGGQ Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 73%.
Acts at least in part as component of the WASH core complex whose assembly at the surface of endosomes inhibits WASH nucleation-promoting factor (NPF) activity in recruiting and activating the Arp2/3 complex to induce actin polymerization and is involved in the fission of tubules that serve as transport intermediates during endosome sorting. Mediates the recruitment of the WASH core complex to endosome membranes via binding to phospholipids and VPS35 of the retromer CSC. Mediates the recruitment of the F-actin-capping protein dimer to the WASH core complex probably promoting localized F-actin polymerization needed for vesicle scission. Via its C-terminus binds various phospholipids, most strongly phosphatidylinositol 4-phosphate (PtdIns-(4)P), phosphatidylinositol 5-phosphate (PtdIns-(5)P) and phosphatidylinositol 3,5-bisphosphate (PtdIns-(3,5)P2). Involved in the endosome-to-plasma membrane trafficking and recycling of SNX27-retromer-dependent cargo proteins, such as GLUT1. Required for the association of DNAJC13, ENTR1, ANKRD50 with retromer CSC subunit VPS35. Required for the endosomal recruitment of CCC complex subunits COMMD1 and CCDC93 as well as the retriever complex subunit VPS35L. [UniProt].
Especificaciones
| FAM21A | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q641Q2 | |
| WASHC2A | |
| Recombinant Protein corresonding to Human FAM21A. Recombinant protein control fragment (Product #RP-108450). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| WASHC2A | |
| alternative protein FAM21B; bA56A21.1; bA98I6.1; FAM21A; FAM21B; family with sequence similarity 21 member A; family with sequence similarity 21, member A; family with sequence similarity 21, member B; WASH complex subunit 2A; WASH complex subunit FAM21A; WASH complex subunit FAM21B; WASHC2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387680 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido