Descripción
The FAM114A1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human FAM114A1. This antibody recognizes human, rat, mouse antigen. The FAM114A1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
Especificaciones
| Antígeno | FAM114A1 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
| Clasificación | Monoclonal |
| Clon | 1B11D6 |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 20 μL |
| Primario o secundario | Primary |
| ID de gen (Entrez) | 92689 |
| Especies diana | Human, Mouse, Rat |
| Mostrar más |