Descripción
Flask certified as sterile, non-cytotoxic and non-pyrogenic.
- Routine performance testing assures consistent culture conditions for reproducible results
- Pull-strip packaging enables easy opening of cases (no knife required)
- Reseal tab on packaging for convenient storage of unused flasks
- Certified for use in the Automation Partnership's SelecT and CompacT SelecT automated cell culture systems
- Straight neck and vented cap
- Totale Volume: 750mL (25.36 oz.)
- Working Volume: 45 to 55mL (1.52 to 1.86 oz)
- Polystyrene
Especificaciones
Especificaciones
| Código de barras | Yes |
| Capacidad (imperial) | 25,3 oz |
| Color del tapón | Azul |
| Material del cierre | Fenólico |
| Capacidad (métrico) | 750 mL |
| Material | Polystyrene |
| Estilo de cuello | Straight |
| Forma | Rectangular |
| Revestimiento de superficie | Tissue Culture Treated |
| Esterilidad | Estéril |
| Mostrar más |