Learn More
Invitrogen™ Factor VIII Monoclonal Antibody (F8-2.2.32), Invitrogen™
Código de producto. 17830356
Change view
Click to view available options
Cantidad:
1 mg
100 μg
500 μL
Tamaño de la unidad:
1 miligramo
100 microgramos
17830356 1 mg 1 miligramo
17860196 100 μg 100 microgramos
11598872 500 μL 1 miligramo
3 options
Código de producto. 17830356
Marca: Invitrogen™ MA545982
Mouse Monoclonal Antibody
MA1-10588 detects full-length human Factor VIII. This antibody does not cross-react with von Willebrand factor. MA1-10588 has been successfully used in ELISA and Western blot applications. The MA1-10588 immunogen is purified human Factor VIII.
This gene encodes coagulation factor VIII, which participates in the intrinsic pathway of blood coagulation; factor VIII is a cofactor for factor IXa which, in the presence of Ca+2 and phospholipids, converts factor X to the activated form Xa. This gene produces two alternatively spliced transcripts. Transcript variant 1 encodes a large glycoprotein, isoform a, which circulates in plasma and associates with von Willebrand factor in a noncovalent complex. This protein undergoes multiple cleavage events. Transcript variant 2 encodes a putative small protein, isoform b, which consists primarily of the phospholipid binding domain of factor VIIIc. This binding domain is essential for coagulant activity. Defects in this gene results in hemophilia A, a common recessive X-linked coagulation disorder.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Factor VIII
Aplicaciones ELISA, Western Blot
Clasificación Monoclonal
Clon F8-2.2.32
Concentración 0.6 mg/mL
Conjugado Unconjugated
Formulación PBS with no preservative; pH 7.4
génica F8
N.º de referencia del gen P00451
Alias de gen AHF; Antihemophilic factor; CF8; Cf-8; coagulation factor VIII; coagulation factor VIII A1 domain; coagulation factor VIII C2 domain; coagulation factor VIII, procoagulant component; coagulation factor VIII, procoagulant component (hemophilia A); coagulation factor VIIIc; DXS1253E; F8; F8B; F8C; Factor 8; Factor VIII; Factor VIII B chain; factor VIII F8B; factor VIII precursor; Factor VIIIa heavy chain, 200 kDa isoform; Factor VIIIa heavy chain, 92 kDa isoform; Factor VIIIa light chain; factor8; FVIII; HEMA; Procoagulant component; RP11-115M6.7
Símbolos de los genes F8
Especie del huésped Mouse
Inmunógeno Purified human Factor VIII.
Método de purificación Protein G
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 2157
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1