missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor V Monoclonal Antibody (5106)

Descripción
MA1-43009 detects Factor V in bovine samples. MA1-43009 has successfully been used in Western blot and ELISA applications. MA1-43009 was derived from bovine factor Va heavy chain.
Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene can result various diseases including Factor V deficiency, thrombophilia due to activated protein C resistance, Budd-Chiari syndrome, Ischemic stroke, and recurrent pregnancy loss.
Especificaciones
Especificaciones
| Antígeno | Factor V |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Monoclonal |
| Clon | 5106 |
| Conjugado | Unconjugated |
| Formulación | 50% water with 50% glycerol and no preservative |
| génica | F5 |
| N.º de referencia del gen | Q28107 |
| Alias de gen | Ac2-120; Activated protein C cofactor; AI173222; Cf5; Cf-5; coagulation factor V; coagulation factor V (proaccelerin, labile factor); Coagulation factor V heavy chain; coagulation factor V jinjiang A2 domain; Coagulation factor V light chain; F5; Factor 5; factor V; factor V Leiden; Factor5; FVL; PCCF; Proaccelerin, labile factor; RPRGL1; THPH2 |
| Símbolos de los genes | F5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?