missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FABP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA552953
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: AFTGKFEMES EKNYDEFMKL LGISSDVIEK ARNFKIVTEV QQDGQDFTWS QHYSGGHTMT NKFTVGKESN IQTMGGKMFK ATVQMEGGKL VVNFPNYHQT SEIVGDKLVE VSTIGGVTYE RVSKRLA Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.
This gene encodes the ileal fatty acid binding protein. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP6 and FABP1 (the liver fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. Transcript variants generated by alternate transcription promoters and/or alternate splicing have been found for this gene.
Especificaciones
| FABP6 | |
| Polyclonal | |
| Unconjugated | |
| FABP6 | |
| FABP6; fatty acid binding protein 6; fatty acid binding protein 6, ileal; fatty acid binding protein 6, ileal (gastrotropin); fatty acid-binding protein 6; Gastrotropin; GT; I-15P; I-BABP; I-BALB; I-BAP; ILBP; ILBP3; ileal bile acid binding protein; ileal bile acid-binding protein; ileal lipid-binding protein; Illbp; illeal lipid-binding protein; intestinal 15 kDa protein; Intestinal bile acid-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2172 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51161 | |
| FABP6 | |
| Recombinant protein corresponding to Human FABP6. Recombinant protein control fragment (Product #RP-89668). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido