missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FABP3/H-FABP Antibody (FABP3/8343R), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-21013AF750
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FABP3/H-FABP Monoclonal antibody specifically detects FABP3/H-FABP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
| FABP3/H-FABP | |
| Monoclonal | |
| Alexa Fluor 750 | |
| FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP | |
| Human recombinant FABP3/H-FABP protein fragment (around aa1-127) (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Cycle and Replication, Cholesterol Metabolism, Lipid and Metabolism | |
| 2170 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FABP3/8343R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido