missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FABP10A Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA572185
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
FABP10A Polyclonal Antibody for Western Blot
Binds hydrophobic ligands, such as cholate, in the cytoplasm. May be involved in intracellular lipid transport. Binds one cholate per subunit.
Especificaciones
| FABP10A | |
| Polyclonal | |
| Unconjugated | |
| fabp10a | |
| fabp10; fabp10a; fatty acid binding protein 10, liver basic; fatty acid binding protein 10a, liver basic; fatty acid-binding protein 10; fatty acid-binding protein 10-A, liver basic; Fatty acid-binding protein, liver; L-BABP; Lb-FABP; Lfabp; L-FABP; liver bile acid-binding protein; liver-basic fatty acid binding protein; liver-type FABP; liver-type fatty acid binding protein; Liver-type fatty acid-binding protein; small cytosolic protein involved in lipid transport and metabolism; Zf-FABP10; zf-Lb-FABP; zgc:103719; zgc:92741; z-L-BABP | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 171481 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9I8L5 | |
| fabp10a | |
| KLH conjugated synthetic peptide between 30-63 amino acids of DANRE fabp10a. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido