Learn More
F4/80 Rat, anti-Mouse, PE, Clone: BM8, STEMCELL Technologies
Rat Monoclonal Antibody
Marca:  Stemcell Technologies 60027PE.1
Encompass_Preferred
 Ver más versiones de este producto
Código de producto. 17178591
  • 155.25€ / 25 microgramos
Descripción
  • The BM8 antibody reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human), an ∼160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors
  • F4/80 is considered a marker of choice for the identification of mature tissue macrophages, being broadly but variably expressed by this cell type in the liver (Kupffer cells), skin (Langerhans cells), bone marrow stroma, pancreas, thymus, spleen (red pulp), lung, and other tissues
  • It is also expressed by circulating monocytes, eosinophils and a subset of dendritic cells
  • F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas
  • This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) and EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770)
Especificaciones
Especificaciones
F4/80
Monoclonal
PE
25 μg
Human, Mouse
Monocytes, Other
Immunohistochemistry, Western Blot, ELISA, Flow Cytometry, Immunocytochemistry
BM8
Rat
Primary
IgG2a κ
Certificados
Promociones
Promociones
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.