Learn More
R&D Systems™ Recombinant F. meningosepticum PNGase F Protein, CF
Effective removal of N-linked oligosaccharides from glycoproteins. Glycerol-free, suitable for downstream MS and HPLC. Get 50% off! Use Promo Code: PNGaseF50 Terms and Conditions
Marca:  R&D Systems™ 9109-GH-020
Encompass
Código de producto. 15740394
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos »
Description
Description
Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10 μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.
Spécification
Spécification
P21163
M.W. (Observed): 34 kDa, reducing conditions; M.W. (theoretical): 36 kDa
E. coli-derived f. meningosepticum PNGase F protein Ala41-Asn354 with N-terminal Met and 6-His tag
<1.0 EU / 1 µg of the protein by the LAL method.
Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.
Yes
Supplied as a 0.2 µm filtered solution in Tris and NaCl.
20 µg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
PNGase F
Unconjugated
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Product Certifications
Promotions
Promotions
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.