missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Rabbit anti-Rat IgG (H+L) Secondary Antibody, HRP
F(ab')2-Rabbit Polyclonal Secondary Antibody
Marca: Invitrogen™ PA129927
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PA1-29927 detects Rat IgG (H+L) from rat samples.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Especificaciones
| Rat IgG (H+L) | |
| Polyclonal | |
| HRP | |
| Rabbit | |
| IgG fraction | |
| RUO | |
| Rat | |
| Secondary Antibody | |
| IgG F(ab')2 |
| ELISA, Immunohistochemistry, Immunomicroscopy, Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2 | |
| Rat IgG whole molecule. | |
| 250 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido