Learn More
Invitrogen™ F(ab')2-Donkey anti-Sheep IgG (H+L) Secondary Antibody, FITC
Código de producto. 15460974
Change view
Click to view available options
Cantidad:
500 μg
Tamaño de la unidad:
0.50 miligramo
15460974 500 μg 0.50 miligramo
1 options
Código de producto. 15460974 Proveedor Invitrogen™ N.º de proveedor A24562
F(ab')2-Donkey Polyclonal Secondary Antibody
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Anti-Sheep secondary antibodies are affinity-purified antibodies with well-characterized specificity for ovine (sheep) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Sheep IgG (H+L)
Aplicaciones Flow Cytometry, Immunocytochemistry
Clasificación Polyclonal
Concentración 1.5 mg/mL
Conjugado FITC
Formulación PBS with 1% BSA and 0.05% sodium azide; pH 7.8
Especie del huésped Donkey
Inmunógeno Gamma Immunoglobins Heavy and Light chains.
Método de purificación Affinity Chromatography
Cantidad 500 μg
Estado normativo RUO
Primario o secundario Secondary
Especies diana Sheep
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Tipo de producto Secondary Antibody
Formulario Liquid
Isotype IgG