missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, AP
Descripción
F(ab')2-Donkey anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody for ELISA, IHC, Western Blot
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Especificaciones
Especificaciones
| Antígeno | Mouse IgG (H+L) Highly Cross-Adsorbed |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1.5 mg/mL |
| Conjugado | Alkaline Phosphatase (AP) |
| Formulación | Secondary antibody with 1% BSA, 30mM triethanolamine, 5mM MgCl2, 0.1mM ZnCl2 and 0.05% sodium azide; pH 7.2 |
| Especie del huésped | Donkey |
| Inmunógeno | Gamma Immunoglobins Heavy and Light chains. |
| Método de purificación | Affinity Chromatography |
| Cantidad | 500 μg |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?