Learn More
Biochrom EZ Read LED Microplate Reader
Descripción
The EZ Read LED microplate reader delivers fast and reliable measurement performance using LEDs coupled to high quality interference filters for light source and wavelength selection.
Novel LED technology provides worry-free operation without compromising on measurement performance.
THe EZ Read LED is PC-controlled using new, intuitive Galapagos software for data acquisition and analysis.
A great choice for busy ELISA labs, EZ Read LED is also available as a UV version for measurements at 340 nm.
EZ Read LED Microplate Reader features:
- Measures one 96-well plate in 5 seconds
- Automatic calibration adn reference channel ensure measure performance
- Measures flat, u-bottom, v-bottom 96-well plates
EZ Read LED Microplate Reader comes in two models:
- Biochrom EZ Read LED
- Biochrom EZ Read LED UV
- ELISA Assay Substrate
- NPP, ABTS use Filter 405 nm
- OPD use Filter 490 nm
- Slow TMB, Turbo TMB, Ultra TMB use Filter 450 nm
- Reference wavelength use Filter 620 nm
- A LED at 340nm can be opted for measurement of NADH/NADPH assays
Galapagos for EZ Read Software's 'click and drag' approach makes usability a key feature.
- Create new plate layouts
- Add multiple assays to a plate
- Add multiple plates per analysis
- Easy to create new methods
- Select only the data you want for data reporting
- Windows 7 & 8 compatibility as well as older Windows OS from XP
- So easy you won't need to export your data into Excel
Especificaciones
Especificaciones
| Tipo de detector | 8 Silicon Photodiodes |
| Fuente de luz | LED |
| Velocidad de medición | 5 sec. |
| Tipos de placas | 96-well Plates |
| Agitación | Estándar de 4 velocidades |
| Software Type | Software Galapagos |
| Selección de longitud de onda | LED |
| Exactitud | ≤ 1% and 0.005 to 3.5 OD |
| Requisitos eléctricos | 24 VDC, 2.5 A |
| Filtros | 405, 450, 492, 620 nm LED |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.