Learn More
EXOG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16821115 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
16821115 150 μL 150 microlitros
16831115 20 μL 20 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 16821115 Proveedor Proteintech N.º de proveedor 215231AP150UL
Rabbit Polyclonal Antibody
This gene encodes an endo/exonuclease with 5′-3′ exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5′ end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.
Especificaciones
Antígeno EXOG
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.23 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica EXOG
N.º de referencia del gen Q8C163, Q9Y2C4
Alias de gen Endo G like 1, ENDOGL1, ENDOGL2, Endonuclease G like 1, ENGL, ENGL a, ENGL B, EXOG, Nuclease EXOG, mitochondrial
Símbolos de los genes EXOG
Especie del huésped Rabbit
Inmunógeno EXOG Fusion Protein Ag16122
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 208194, 301062, 9941
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG